Anti-CD84 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CD84.1.21]

Supplier: GeneTex
GTX75850
89278-330EA 543.95 USD
89278-330
Anti-CD84 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CD84.1.21]
Antibodies
Mouse monoclonal antibody [CD84.1.21] to CD84

Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: CD84.1.21
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR