Anti-CD84 Mouse Monoclonal Antibody [clone: CD84.1.21]

Supplier: GeneTex
GTX75849
89279-938EA 631.47 USD
89279-938
Anti-CD84 Mouse Monoclonal Antibody [clone: CD84.1.21]
Antibodies
Mouse monoclonal antibody [CD84.1.21] to CD84

Recommended Starting Dilutions: Flow Cytometry: 1/100 - 1/200 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD84
Clonality: Monoclonal
Clone: CD84.1.21
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR