Anti-CD86 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MRC OX-48]

GTX75848
89265-604EA 471.29 USD
89265-604
Anti-CD86 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MRC OX-48]
Mouse monoclonal antibody [OX-48] to CD86

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD86
Clonality: Monoclonal
Clone: MRC OX-48
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR