Anti-CD46 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MEM-258]

Supplier: GeneTex
GTX75835
89261-392EA 526.79 USD
89261-392
Anti-CD46 Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: MEM-258]
Antibodies
Mouse monoclonal antibody [MEM-258] to CD46

Type: Primary
Antigen: CD46
Clonality: Monoclonal
Clone: MEM-258
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR