Anti-BIRC8 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX59774
89283-496EA 552.71 USD
89283-496
Anti-BIRC8 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ILP-2

Western blotting (0.5-4 µg/ml). However, the optimal dilution conditions should be determined individually. The affinity-purified rabbit antibody recognizes ~32 kDa ILP-2 in samples from human, mouse and rat origins.

Type: Primary
Antigen: BIRC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR