Anti-EIF5A Mouse Monoclonal Antibody [clone: AT2E9]

Supplier: GeneTex
GTX50006
89298-510EA 512.17 USD
89298-510
Anti-EIF5A Mouse Monoclonal Antibody [clone: AT2E9]
Antibodies
Mouse monoclonal antibody [AT2E9] to EIF5A

Type: Primary
Antigen: EIF5A
Clonality: Monoclonal
Clone: AT2E9
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR