Anti-TTR Antibody Pair

Supplier: GeneTex
GTX500029
89333-274EA 618.81 USD
89333-274
Anti-TTR Antibody Pair
Antibodies
Validated pair of antibodies for Transthyretin

We recommend using the capture antibody at a concentration of 5µg/mL and the detection antibody at a concentration of 5µg/mL. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: TTR
Clonality: 0
Clone:
Conjugation:
Epitope:
Host: Multiple
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR