Anti-SUMO2 Mouse Monoclonal Antibody [clone: AT10F1]

Supplier: GeneTex
GTX50000
89336-028EA 614.81 USD
89336-028
Anti-SUMO2 Mouse Monoclonal Antibody [clone: AT10F1]
Antibodies
Mouse monoclonal antibody [AT10F1] to SUMO2

Type: Primary
Antigen: SUMO2
Clonality: Monoclonal
Clone: AT10F1
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR