Anti-SPANXC Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48817
89333-552EA 660.25 USD
89333-552
Anti-SPANXC Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to SPANXC

Recommended Starting Dilutions: WB: Use at 1:1,000 - 1:5,000 ELISA: Use at 1:5,000 - 1:20,000 IHC: Use at 1.25-2.5 ug/ml

Type: Primary
Antigen: SPANXC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR