Anti-TMBIM1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48815
89317-726EA 606.86 USD
89317-726
Anti-TMBIM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to Tmbim1

Recommended Starting Dilutions: WB: Use at 1:500 - 1:2,000 ELISA: Use at 1:10,000

Type: Primary
Antigen: TMBIM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR