Anti-VEGFA Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX48811
89317-718EA 606.86 USD
89317-718
Anti-VEGFA Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to VEGF

Recommended Starting Dilutions: WB: Use at 1:333 ELISA: Use at User Optimized

Type: Primary
Antigen: VEGFA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR