Anti-AQP11 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX47927
89279-810EA 602.76 USD
89279-810
Anti-AQP11 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Aquaporin 11

We recommend the following starting dilutions: 1:500 for WB 1:150 for IHC Optimal working dilutions should be determined experimentally be the end user.

Type: Primary
Antigen: AQP11
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR