Anti-PGAP3 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX47869
89279-738EA 575.18 USD
89279-738
Anti-PGAP3 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to Post-GPI attachment to proteins 3 (C-terminal)

We recommend the following starting dilutions: 1:500 for WB 1:150 for IHC Optimal working dilutions should be determined experimentally be the end user.

Type: Primary
Antigen: PGAP3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR