Anti-CD317 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX47796
89279-662EA 562.02 USD
89279-662
Anti-CD317 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CD317

We recommend the following starting dilutions: WB:1:500. ELISA:1:10,000. Optimal working dilutions should be determined experimentally be the end user.

Type: Primary
Antigen: CD317
Clonality: Polyclonal
Clone: 0
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR