Anti-PHACTR1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX45287
89267-034EA 597.48 USD
89267-034
Anti-PHACTR1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PHACTR1 (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: PHACTR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR