Anti-C10ORF30 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX45264
89266-988EA 612.19 USD
89266-988
Anti-C10ORF30 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to C10orf30 (N-terminal)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: C10ORF30
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR