Anti-BEND7 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX45263
89266-986EA 612.19 USD
89266-986
Anti-BEND7 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to BEND7 (Middle)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: BEND7
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR