Anti-ZDHHC21 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX45197
89266-858EA 627.59 USD
89266-858
Anti-ZDHHC21 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to ZDHHC21 (Middle)

Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: ZDHHC21
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR