Anti-LRRTM1 Rabbit Polyclonal Antibody

Supplier: GeneTex
GTX45194
89266-852EA 612.19 USD
89266-852
Anti-LRRTM1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to LRRTM1 (Middle)

Recommended Dilutions: For Western Blot: Use at 1 ug/ml.<p>Not yet tested in other applications. Optimal dilutions should be determined experimentally by the researcher.

Type: Primary
Antigen: LRRTM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR