Anti-EC 3.5.2.8 Mouse Monoclonal Antibody [clone: 8A5.A10]

Supplier: GeneTex
GTX44346
89279-640EA 503.03 USD
89279-640
Anti-EC 3.5.2.8 Mouse Monoclonal Antibody [clone: 8A5.A10]
Antibodies
Type: Primary
Antigen: EC 3.5.2.8
Clonality: Monoclonal
Clone: 8A5.A10
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR