Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: YKIX716.13]

Supplier: GeneTex
GTX43583
89262-990EA 505.13 USD
89262-990
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: YKIX716.13]
Antibodies
Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: YKIX716.13
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Dog
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR