Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: YW62.3]

GTX43581
89307-322EA 199.63 USD
89307-322
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: YW62.3]
Rat monoclonal antibody [YW62.3] to CD45

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: YW62.3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR