Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu78]

Supplier: GeneTex
GTX43576
89279-572EA 463.48 USD
89279-572
Anti-PTPRC Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu78]
Mouse monoclonal antibody [Bu78] to CD45RA

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: Bu78
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR