Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-19]

GTX43545
89333-528EA 377.04 USD
89333-528
Anti-CD5 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-19]
Mouse monoclonal antibody [OX-19] to CD5

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: OX-19
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR