Anti-CD5 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: YKIX322.3]

Supplier: GeneTex
GTX43543
89262-974EA 505.13 USD
89262-974
Anti-CD5 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: YKIX322.3]
Antibodies
Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: YKIX322.3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR