Anti-ICAM3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu65]

GTX43540
89279-566EA 442.18 USD
89279-566
Anti-ICAM3 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu65]
Mouse monoclonal antibody [Bu65] to ICAM3

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: Bu65
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR