Anti-CD55 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu14]

Supplier: GeneTex
GTX43536
89279-564EA 655.63 USD
89279-564
Anti-CD55 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu14]
Antibodies
Mouse monoclonal antibody [Bu14] to CD55

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: Bu14
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR