Anti-ITGB3 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HM beta 3.1]

GTX43531
89281-116EA 511.79 USD
89281-116
Anti-ITGB3 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: HM beta 3.1]
Hamster monoclonal antibody [HM beta 3.1] to Integrin beta 3

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: ITGB3
Clonality: Monoclonal
Clone: HM beta 3.1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR