Anti-CD68 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: FA-11]

Supplier: GeneTex
GTX43518
89315-058EA 324.95 USD
89315-058
Anti-CD68 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: FA-11]
Antibodies
Rat monoclonal antibody [FA-11] to CD68

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD68
Clonality: Monoclonal
Clone: FA-11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR