Anti-CD8 Alpha Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-8]

Supplier: GeneTex
GTX43506
89292-730EA 336.12 USD
89292-730
Anti-CD8 Alpha Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-8]
Antibodies
Mouse monoclonal antibody [OX-8] to CD8 alpha

Type: Primary
Antigen: CD8 Alpha
Clonality: Monoclonal
Clone: OX-8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR