Anti-CD8 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu88]

Supplier: GeneTex
GTX43501
89279-556EA 550.31 USD
89279-556
Anti-CD8 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Bu88]
Antibodies
Mouse monoclonal antibody [Bu88] to CD8

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD8
Clonality: Monoclonal
Clone: Bu88
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR