Anti-CD81 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Eat2]

Supplier: GeneTex
GTX43491
89292-724EA 567.5 USD
89292-724
Anti-CD81 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: Eat2]
Antibodies
Hamster monoclonal antibody [Eat2] to CD81

Type: Primary
Antigen: CD81
Clonality: Monoclonal
Clone: Eat2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR