Anti-CD83 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3D11]

Supplier: GeneTex
GTX43489
89292-722EA 652.21 USD
89292-722
Anti-CD83 Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 3D11]
Antibodies
Rat monoclonal antibody [3D11] to CD83

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CD83
Clonality: Monoclonal
Clone: 3D11
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR