Anti-CTL1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: VIM-15b]

Supplier: GeneTex
GTX43476
89279-548EA 534.27 USD
89279-548
Anti-CTL1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: VIM-15b]
Antibodies
Mouse monoclonal antibody [VIM-15b] to CD92

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/5 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: CTL1
Clonality: Monoclonal
Clone: VIM-15b
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR