Anti-IFNG Mouse Monoclonal Antibody (FITC) [clone: CC302]

Supplier: GeneTex
GTX43430
89262-954EA 903.01 USD
89262-954
Anti-IFNG Mouse Monoclonal Antibody (FITC) [clone: CC302]
Antibodies
Mouse monoclonal antibody [CC302] to IFN gamma

Recommended Starting Dilutions: Flow Cytometry: Max Dilution 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: IFNG
Clonality: Monoclonal
Clone: CC302
Conjugation: FITC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Dog, Pig, Dolphin, Ferret, Goat, Horse, Mink, Sheep, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR