Anti-KLRG1 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2F1]

Supplier: GeneTex
GTX43425
89292-714EA 489.94 USD
89292-714
Anti-KLRG1 Hamster Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 2F1]
Antibodies
Hamster monoclonal antibody [2F1] to KLRG1

Type: Primary
Antigen: KLRG1
Clonality: Monoclonal
Clone: 2F1
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Hamster
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR