Anti-IgL Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: GeneTex
GTX43421
89277-534EA 505.24 USD
89277-534
Anti-IgL Goat Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
FITC-conjugated Goat Polyclonal antibody to Human lambda Light chain

Recommended Starting Dilutions: For FACS: Use Neat - 1/10 For For ICC/IF: Use Neat - 1/10 Optimal dilutions should be determined experimentally by the researcher.

Type: Secondary
Antigen: IgL
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR