Anti-Ly-6B.2 AlloAntigen Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 39266]

Supplier: GeneTex
GTX43417
89285-702EA 457.74 USD
89285-702
Anti-Ly-6B.2 AlloAntigen Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 39266]
Antibodies
Type: Primary
Antigen: Ly-6B.2 AlloAntigen
Clonality: Monoclonal
Clone: 39266
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR