Anti-MHC Class II DR Monomorphic Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 37.68]

Supplier: GeneTex
GTX43393
89262-942EA 507.33 USD
89262-942
Anti-MHC Class II DR Monomorphic Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 37.68]
Antibodies
Type: Primary
Antigen: MHC Class II DR Monomorphic
Clonality: Monoclonal
Clone: 37.68
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Bovine, Goat, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR