Anti-MHC Class II H-2I-Ab/S Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-3]

Supplier: GeneTex
GTX43392
89303-534EA 604.12 USD
89303-534
Anti-MHC Class II H-2I-Ab/S Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: OX-3]
Antibodies
Type: Primary
Antigen: MHC Class II H-2I-Ab/S
Clonality: Monoclonal
Clone: OX-3
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR