Anti-MHC Class II Monomorphic Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 21-1A6]

Supplier: GeneTex
GTX43388
89262-940EA 507.33 USD
89262-940
Anti-MHC Class II Monomorphic Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 21-1A6]
Antibodies
Type: Primary
Antigen: MHC Class II Monomorphic
Clonality: Monoclonal
Clone: 21-1A6
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Chicken
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR