Anti-MHC Class II Rt1B Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: F17-23-2]

Supplier: GeneTex
GTX43386
89285-696EA 540.8 USD
89285-696
Anti-MHC Class II Rt1B Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: F17-23-2]
Antibodies
Type: Primary
Antigen: MHC Class II Rt1B
Clonality: Monoclonal
Clone: F17-23-2
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR