Anti-Nkg2A/C/E Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 20d5]

GTX43377
89281-110EA 523.61 USD
89281-110
Anti-Nkg2A/C/E Rat Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 20d5]
Type: Primary
Antigen: Nkg2A/C/E
Clonality: Monoclonal
Clone: 20d5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype:
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR