Anti-CD11A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BL1H8]

Supplier: GeneTex
GTX43371
89262-936EA 919.76 USD
89262-936
Anti-CD11A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BL1H8]
Antibodies
Type: Primary
Antigen: CD11A
Clonality: Monoclonal
Clone: BL1H8
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Pig, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR