Anti-CD172A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BL1H7]

Supplier: GeneTex
GTX43365
89262-926EA 873.33 USD
89262-926
Anti-CD172A Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: BL1H7]
Antibodies
Type: Primary
Antigen: CD172A
Clonality: Monoclonal
Clone: BL1H7
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR