Anti-T Lymphocytes Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: KEN-5]

Supplier: GeneTex
GTX43339
89265-454EA 507.33 USD
89265-454
Anti-T Lymphocytes Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: KEN-5]
Antibodies
Type: Primary
Antigen: T Lymphocytes
Clonality: Monoclonal
Clone: KEN-5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR