Anti-V5 Tag Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SV5-Pk5]

Supplier: GeneTex
GTX43333
89266-060EA 595.88 USD
89266-060
Anti-V5 Tag Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SV5-Pk5]
Antibodies
Type: Primary
Antigen: V5 Tag
Clonality: Monoclonal
Clone: SV5-Pk5
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR