Anti-Wc1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CC101]

Supplier: GeneTex
GTX43330
89262-896EA 507.33 USD
89262-896
Anti-Wc1 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: CC101]
Antibodies
Mouse monoclonal antibody [CC101] to Wc1

Recommended Starting Dilutions: Flow Cytometry: Neat - 1/10 Not tested in other applications. Optimal dilution / concentration should be determined by the end user.

Type: Primary
Antigen: Wc1
Clonality: Monoclonal
Clone: CC101
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Bovine, Pig
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR