Anti-ZAP 70 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SBZAP]

GTX43323
89283-142EA 442.34 USD
89283-142
Anti-ZAP 70 Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SBZAP]
Mouse monoclonal antibody [SBZAP] to ZAP70

Type: Primary
Antigen: ZAP 70
Clonality: Monoclonal
Clone: SBZAP
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR