Anti-Defensin Mouse Monoclonal Antibody [clone: B539M]

GTX43315
89299-806EA 341.41 USD
89299-806
Anti-Defensin Mouse Monoclonal Antibody [clone: B539M]
Type: Primary
Antigen: Defensin
Clonality: Monoclonal
Clone: B539M
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR